Assay Details
| Gene Symbol: Hexa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010421.4 | NP_034551.2 | 6 - 7 | 725 | 79 | |
| GenBank mRNA | AK075895.1 | - | 6 - 7 | 701 | 79 | |
| AK075911.1 | - | 6 - 7 | 726 | 79 | ||
| AK080777.1 | - | 6 - 7 | 698 | 79 | ||
| AK144168.1 | - | 6 - 7 | 853 | 79 | ||
| AK150444.1 | - | 6 - 7 | 696 | 79 | ||
| AK151960.1 | - | 6 - 7 | 700 | 79 | ||
| AK159814.1 | - | 6 - 7 | 711 | 79 | ||
| AK159890.1 | - | 6 - 7 | 698 | 79 | ||
| AK160112.1 | - | 6 - 7 | 685 | 79 | ||
| AK167013.1 | - | 6 - 7 | 711 | 79 | ||
| AK168186.1 | - | 6 - 7 | 725 | 79 | ||
| AK170903.1 | - | 6 - 7 | 712 | 79 | ||
| AK179213.1 | - | 1 - 2 | 72 | 79 | ||
| AK218113.1 | - | 1 - 2 | 72 | 79 | ||
| BC010755.1 | - | 6 - 7 | 694 | 4225233 | 79 | |
| U07631.1 | - | 6 - 7 | 717 | 79 | ||
| X64331.1 | - | 6 - 7 | 668 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hexa
Genomic Map