Assay Details
| Gene Symbol: Hexa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010421.4 | NP_034551.2 | 9 - 10 | 1126 | 63 | |
| GenBank mRNA | AK075895.1 | - | 9 - 10 | 1102 | 63 | |
| AK075911.1 | - | 9 - 10 | 1127 | 63 | ||
| AK080777.1 | - | 9 - 10 | 1099 | 63 | ||
| AK144168.1 | - | 9 - 10 | 1254 | 63 | ||
| AK150444.1 | - | 9 - 10 | 1097 | 63 | ||
| AK151960.1 | - | 9 - 10 | 1101 | 63 | ||
| AK159091.1 | - | 10 - 11 | 1097 | 63 | ||
| AK159814.1 | - | 9 - 10 | 1112 | 63 | ||
| AK160112.1 | - | 9 - 10 | 1086 | 63 | ||
| AK167013.1 | - | 9 - 10 | 1112 | 63 | ||
| AK168186.1 | - | 9 - 10 | 1126 | 63 | ||
| AK170903.1 | - | 9 - 10 | 1113 | 63 | ||
| AK213356.1 | - | 1 - 2 | 81 | 63 | ||
| BC010755.1 | - | 9 - 10 | 1095 | 4225233 | 63 | |
| U07631.1 | - | 9 - 10 | 1118 | 63 | ||
| X64331.1 | - | 9 - 10 | 1069 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hexa
Genomic Map