Assay Details
| Gene Symbol: Ifngr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010511.2 | NP_034641.1 | 3 - 4 | 517 | 81 | |
| GenBank mRNA | AF128216.1 | - | 3 - 4 | 468 | 81 | |
| AF128217.1 | - | 3 - 4 | 468 | 81 | ||
| AK150503.1 | - | 3 - 4 | 466 | 81 | ||
| AK151079.1 | - | 3 - 4 | 471 | 81 | ||
| AK152085.1 | - | 3 - 4 | 466 | 81 | ||
| AK152384.1 | - | 3 - 4 | 466 | 81 | ||
| AK153084.1 | - | 3 - 4 | 466 | 81 | ||
| AK153347.1 | - | 3 - 4 | 469 | 81 | ||
| AK159359.1 | - | 3 - 4 | 471 | 81 | ||
| AK159660.1 | - | 3 - 4 | 411 | 81 | ||
| AK159839.1 | - | 3 - 4 | 411 | 81 | ||
| AK169593.1 | - | 3 - 4 | 486 | 81 | ||
| AY157836.1 | - | 3 - 4 | 468 | 81 | ||
| AY157837.1 | - | 3 - 4 | 468 | 81 | ||
| BC132599.1 | - | 3 - 4 | 418 | 40130876 | 81 | |
| BC138617.1 | - | 3 - 4 | 418 | 8861637 | 81 | |
| J05265.1 | - | 3 - 4 | 494 | 81 | ||
| M25764.1 | - | 3 - 4 | 419 | 81 | ||
| M26711.1 | - | 3 - 4 | 494 | 81 | ||
| M28233.1 | - | 3 - 4 | 490 | 81 | ||
| M28995.1 | - | 3 - 4 | 395 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifngr1
Genomic Map