Assay Details
| Gene Symbol: Plod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011122.3 | NP_035252.1 | 13 - 14 | 1578 | 61 | |
| GenBank mRNA | AF046782.1 | - | 13 - 14 | 1506 | 61 | |
| AK137480.1 | - | 13 - 14 | 1529 | 61 | ||
| BC006599.1 | - | 13 - 14 | 1503 | 3965301 | 61 | |
| BC010268.1 | - | 5 - 6 | 523 | 3589182 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plod1
Genomic Map