| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400033087.mm.1 |
TC0400004004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400033087.mm.1 |
NM_011122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033087.mm.1 |
NM_011122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033095.mm.1 |
TC0400004004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400033095.mm.1 |
NM_011122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033095.mm.1 |
NM_011122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033086.mm.1 |
TC0400004004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400033086.mm.1 |
NM_011122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033086.mm.1 |
NM_011122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033091.mm.1 |
TC0400004004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400033091.mm.1 |
NM_011122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033091.mm.1 |
NM_011122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033088.mm.1 |
TC0400004004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400033088.mm.1 |
NM_011122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033088.mm.1 |
NM_011122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033090.mm.1 |
TC0400004004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400033090.mm.1 |
NM_011122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033090.mm.1 |
NM_011122.3 |
Refseq |