Assay Details
| Gene Symbol: Plod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011122.3 | NP_035252.1 | 4 - 5 | 569 | 70 | |
| GenBank mRNA | AF046782.1 | - | 4 - 5 | 497 | 70 | |
| AK131632.1 | - | 4 - 5 | 518 | 70 | ||
| AK137480.1 | - | 4 - 5 | 519 | 70 | ||
| AK157729.1 | - | 4 - 5 | 519 | 70 | ||
| AK160822.1 | - | 4 - 5 | 536 | 70 | ||
| BC006599.1 | - | 4 - 5 | 494 | 3965301 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plod1
Genomic Map