Assay Details
| Gene Symbol: Plod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011122.3 | NP_035252.1 | 6 - 7 | 745 | 68 | |
| GenBank mRNA | AF046782.1 | - | 6 - 7 | 673 | 68 | |
| AK137480.1 | - | 6 - 7 | 695 | 68 | ||
| AK157729.1 | - | 6 - 7 | 695 | 68 | ||
| AK160822.1 | - | 6 - 7 | 712 | 68 | ||
| AK206290.1 | - | 1 - 2 | 75 | 68 | ||
| BC006599.1 | - | 6 - 7 | 670 | 3965301 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plod1
Genomic Map