Assay Details
| Gene Symbol: Plod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011122.3 | NP_035252.1 | 7 - 8 | 845 | 63 | |
| GenBank mRNA | AF046782.1 | - | 7 - 8 | 773 | 63 | |
| AK137480.1 | - | 7 - 8 | 795 | 63 | ||
| AK157729.1 | - | 7 - 8 | 795 | 63 | ||
| AK160822.1 | - | 7 - 8 | 812 | 63 | ||
| BC006599.1 | - | 7 - 8 | 770 | 3965301 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plod1
Genomic Map