Assay Details
| Gene Symbol: Prlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011169.5 | NP_035299.4 | 9 - 10 | 1585 | 92 | |
| GenBank mRNA | AK054551.1 | - | 8 - 9 | 2096 | 92 | |
| BC005555.1 | - | 9 - 10 | 1168 | 3586308 | 92 | |
| BC006652.1 | - | 9 - 10 | 1059 | 3500076 | 92 | |
| D10214.1 | - | 7 - 8 | 860 | 92 | ||
| L13593.1 | - | 7 - 8 | 895 | 92 | ||
| L14811.1 | - | 7 - 8 | 837 | 92 | ||
| X73372.1 | - | 7 - 8 | 837 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prlr
Genomic Map