| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900009054.mm.1 |
NM_011262.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009054.mm.1 |
TC1900000990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009046.mm.1 |
NM_011262.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009046.mm.1 |
TC1900000990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009047.mm.1 |
NM_011262.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009047.mm.1 |
TC1900000990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009045.mm.1 |
NM_011262.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009045.mm.1 |
TC1900000990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009041.mm.1 |
NM_011262.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009041.mm.1 |
TC1900000990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009049.mm.1 |
NM_011262.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009049.mm.1 |
TC1900000990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009043.mm.1 |
NM_011262.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009043.mm.1 |
TC1900000990.mm.1 |
MTA TC |