Assay Details
| Gene Symbol: Ruvbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011304.3 | NP_035434.1 | 10 - 11 | 1084 | 56 | |
| GenBank mRNA | AB013912.1 | - | 10 - 11 | 915 | 56 | |
| AK135404.1 | - | 10 - 11 | 898 | 56 | ||
| AK144156.1 | - | 10 - 11 | 906 | 56 | ||
| AK159112.1 | - | 10 - 11 | 1086 | 56 | ||
| AK191656.1 | - | 2 - 3 | 140 | 56 | ||
| AK210146.1 | - | 2 - 3 | 140 | 56 | ||
| AK212334.1 | - | 2 - 3 | 140 | 56 | ||
| BC072585.1 | - | 1583 | 56 | |||
| BC115809.1 | - | 10 - 11 | 902 | 40105495 | 56 | |
| BC116693.1 | - | 10 - 11 | 901 | 40105493 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ruvbl2
Genomic Map