Assay Details
| Gene Symbol: Ruvbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011304.3 | NP_035434.1 | 13 - 14 | 1454 | 102 | |
| GenBank mRNA | AB013912.1 | - | 13 - 14 | 1285 | 102 | |
| AK135404.1 | - | 13 - 14 | 1268 | 102 | ||
| AK144156.1 | - | 13 - 14 | 1276 | 102 | ||
| AK159112.1 | - | 13 - 14 | 1456 | 102 | ||
| BC072585.1 | - | 1953 | 102 | |||
| BC115809.1 | - | 13 - 14 | 1272 | 40105495 | 102 | |
| BC116693.1 | - | 13 - 14 | 1271 | 40105493 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ruvbl2
Genomic Map