Assay Details
| Gene Symbol: Ruvbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011304.3 | NP_035434.1 | 4 - 5 | 467 | 101 | |
| GenBank mRNA | AB013912.1 | - | 4 - 5 | 298 | 101 | |
| AK144156.1 | - | 4 - 5 | 289 | 101 | ||
| AK159112.1 | - | 4 - 5 | 469 | 101 | ||
| AK187938.1 | - | 1 - 2 | 56 | 101 | ||
| BC072585.1 | - | 966 | 101 | |||
| BC115809.1 | - | 4 - 5 | 285 | 40105495 | 101 | |
| BC116693.1 | - | 4 - 5 | 284 | 40105493 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ruvbl2
Genomic Map