Assay Details
| Gene Symbol: Spag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012031.3 | NP_036161.2 | 17 - 18 | 2444 | 62 | |
| GenBank mRNA | AF181252.1 | - | 17 - 18 | 2436 | 62 | |
| AF181253.1 | - | 7 - 8 | 1128 | 62 | ||
| AK032601.1 | - | 14 - 15 | 2239 | 62 | ||
| BC046313.1 | - | 17 - 18 | 2444 | 6310604 | 62 | |
| BC098201.1 | - | 1 - 2 | 1774 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spag1
Genomic Map