Assay Details
| Gene Symbol: Spag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012031.3 | NP_036161.2 | 6 - 7 | 841 | 101 | |
| GenBank mRNA | AF181252.1 | - | 6 - 7 | 833 | 101 | |
| AK032601.1 | - | 3 - 4 | 636 | 101 | ||
| BC046313.1 | - | 6 - 7 | 841 | 6310604 | 101 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK032601.1 transcripts.
Set Membership: |
|

Spag1
Genomic Map