Assay Details
| Gene Symbol: Aoah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012054.4 | NP_036184.1 | 16 - 17 | 1581 | 76 | |
| GenBank mRNA | AK084452.1 | - | 16 - 17 | 1576 | 76 | |
| AK085343.1 | - | 16 - 17 | 1553 | 76 | ||
| AK133301.1 | - | 16 - 17 | 1522 | 76 | ||
| AK171572.1 | - | 16 - 17 | 1400 | 76 | ||
| CT010242.1 | - | 16 - 17 | 1300 | 76 | ||
More Information
Set Membership: |
|

Aoah
Genomic Map