Assay Details
Gene Symbol: Gfpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013528.3 | NP_038556.1 | 3 - 4 | 374 | 118 | |
GenBank mRNA | AF334736.1 | - | 3 - 4 | 331 | 118 | |
AK011010.1 | - | 586 | 118 | |||
AK019852.1 | - | 3 - 4 | 370 | 118 | ||
AK039792.1 | - | 4 - 5 | 431 | 118 | ||
AK087173.1 | - | 3 - 4 | 375 | 118 | ||
BB664274.1 | - | 3 - 4 | 375 | 118 | ||
BC010516.1 | - | 3 - 4 | 322 | 4191553 | 118 | |
BC050762.1 | - | 3 - 4 | 370 | 6742987 | 118 | |
U00932.1 | - | 2 - 3 | 221 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
