Assay Details
| Gene Symbol: P3h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013534.4 | NP_038562.2 | 7 - 8 | 1246 | 67 | |
| GenBank mRNA | AJ441086.1 | - | 7 - 8 | 1246 | 67 | |
| AK184825.1 | - | 33 | 67 | |||
| AY463530.1 | - | 7 - 8 | 1246 | 67 | ||
| BC003726.1 | - | 7 - 8 | 832 | 3497610 | 67 | |
| BC016431.1 | - | 6 - 7 | 732 | 3483280 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

P3h3
Genomic Map