Assay Details
| Gene Symbol: Osbpl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199227.1 | NP_001186156.1 | 4 - 5 | 411 | 60 | |
| NM_024289.2 | NP_077251.1 | 4 - 5 | 611 | 60 | ||
| GenBank mRNA | AB074008.1 | - | 4 - 5 | 404 | 60 | |
| AJ278263.2 | - | 4 - 5 | 611 | 60 | ||
| BC079872.1 | - | 4 - 5 | 392 | 6832073 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl5
Genomic Map