Assay Details
| Gene Symbol: Osbpl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199227.1 | NP_001186156.1 | 9 - 10 | 1168 | 88 | |
| NM_024289.2 | NP_077251.1 | 9 - 10 | 1368 | 88 | ||
| GenBank mRNA | AB074008.1 | - | 9 - 10 | 1161 | 88 | |
| AJ278263.2 | - | 9 - 10 | 1368 | 88 | ||
| BC079872.1 | - | 9 - 10 | 1149 | 6832073 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl5
Genomic Map