Assay Details
| Gene Symbol: Rnf135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028019.3 | NP_082295.1 | 3 - 4 | 682 | 81 | |
| GenBank mRNA | AK010429.1 | - | 3 - 4 | 684 | 81 | |
| AK150921.1 | - | 3 - 4 | 683 | 81 | ||
| AK152096.1 | - | 3 - 4 | 675 | 81 | ||
| AK185113.1 | - | 2 - 3 | 81 | |||
| BC096385.1 | - | 3 - 4 | 676 | 30744704 | 81 | |
| BC132307.1 | - | 3 - 4 | 678 | 40130584 | 81 | |
| BC138303.1 | - | 3 - 4 | 678 | 8861322 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf135
Genomic Map