Assay Details
| Gene Symbol: Tango2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138583.2 | NP_613049.2 | 2 - 3 | 130 | 80 | |
| GenBank mRNA | AK089239.1 | - | 2 - 3 | 112 | 80 | |
| AK133588.1 | - | 2 - 3 | 113 | 80 | ||
| AK168595.1 | - | 1 - 2 | 109 | 80 | ||
| BC005445.1 | - | 2 - 3 | 191 | 3496749 | 80 | |
| X74504.1 | - | 1 - 2 | 103 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tango2
Genomic Map