Assay Details
| Gene Symbol: Zfp282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146175.3 | NP_666287.3 | 2 - 3 | 708 | 108 | |
| GenBank mRNA | AK030516.1 | - | 2 - 3 | 616 | 108 | |
| AK081854.1 | - | 2 - 3 | 607 | 108 | ||
| AK088531.1 | - | 2 - 3 | 707 | 108 | ||
| AK148038.1 | - | 2 - 3 | 694 | 108 | ||
| BC025581.1 | - | 2 - 3 | 610 | 5325029 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp282
Genomic Map