Assay Details
| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK183644.1 | - | 2 - 3 | 289 | 76 | |
| BC036177.1 | - | 14 - 15 | 1854 | 5004341 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dis3l2
Genomic Map