Assay Details
| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172157.1 | NP_001165628.1 | 4 - 5 | 425 | 144 | |
| NM_153530.2 | NP_705758.1 | 4 - 5 | 425 | 144 | ||
| GenBank mRNA | AK029652.1 | - | 4 - 5 | 402 | 144 | |
| AK031180.1 | - | 4 - 5 | 408 | 144 | ||
| AK040613.1 | - | 4 - 5 | 402 | 144 | ||
| BC036177.1 | - | 4 - 5 | 386 | 5004341 | 144 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK029652.1 transcripts.
Set Membership: |
|

Dis3l2
Genomic Map