Assay Details
| Gene Symbol: Zfand6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022985.6 | NP_075361.2 | 3 - 4 | 377 | 128 | |
| GenBank mRNA | AJ251508.1 | - | 283 | 128 | ||
| AK002775.1 | - | 3 - 4 | 378 | 128 | ||
| AK009403.1 | - | 2 - 3 | 235 | 128 | ||
| AK144902.1 | - | 2 - 3 | 227 | 128 | ||
| AK212602.1 | - | 1 - 2 | 104 | 128 | ||
| BC010683.1 | - | 2 - 3 | 238 | 2647905 | 128 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK014007.1 | - | Chr. - - [-] on Build | 128 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Zfand6
Genomic Map