Assay Details
| Gene Symbol: Uba7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023738.4 | NP_076227.1 | 12 - 13 | 1543 | 95 | |
| GenBank mRNA | AF288694.1 | - | 7 - 8 | 805 | 95 | |
| AK004894.2 | - | 12 - 13 | 1544 | 95 | ||
| AK054410.1 | - | 12 - 13 | 1550 | 95 | ||
| AK089235.1 | - | 12 - 13 | 1547 | 95 | ||
| AK129009.1 | - | 12 - 13 | 1453 | 95 | ||
| AK170421.1 | - | 12 - 13 | 1536 | 95 | ||
| AK170644.1 | - | 11 - 12 | 1918 | 95 | ||
| AK172467.1 | - | 11 - 12 | 1881 | 95 | ||
| BC037693.1 | - | 10 - 11 | 1317 | 95 | ||
| BC138843.1 | - | 12 - 13 | 1460 | 8861865 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uba7
Genomic Map