Assay Details
| Gene Symbol: Uba7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023738.4 | NP_076227.1 | 1 - 2 | 78 | ||
| GenBank mRNA | AK004894.2 | - | 1 - 2 | 78 | ||
| AK054410.1 | - | 1 - 2 | 78 | |||
| AK089235.1 | - | 1 - 2 | 78 | |||
| AK089256.1 | - | 1 - 2 | 78 | |||
| AK129009.1 | - | 1 - 2 | 78 | |||
| AK170421.1 | - | 1 - 2 | 78 | |||
| AK170644.1 | - | 1 - 2 | 78 | |||
| AK172467.1 | - | 1 - 2 | 78 | |||
| BC138843.1 | - | 1 - 2 | 8861865 | 78 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK004894.2,AK089235.1,BC138843.1,NM_023738.4 transcripts.
Set Membership: |
|

Uba7
Genomic Map