Assay Details
| Gene Symbol: Uba7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023738.4 | NP_076227.1 | 20 - 21 | 2539 | 66 | |
| GenBank mRNA | AF288694.1 | - | 15 - 16 | 1801 | 66 | |
| AK004894.2 | - | 20 - 21 | 2540 | 66 | ||
| AK054410.1 | - | 19 - 20 | 2631 | 66 | ||
| AK089235.1 | - | 19 - 20 | 2432 | 66 | ||
| AK129009.1 | - | 20 - 21 | 2449 | 66 | ||
| AK170421.1 | - | 19 - 20 | 2617 | 66 | ||
| AK170644.1 | - | 18 - 19 | 2998 | 66 | ||
| BC037693.1 | - | 18 - 19 | 2313 | 66 | ||
| BC138843.1 | - | 20 - 21 | 2456 | 8861865 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uba7
Genomic Map