Assay Details
| Gene Symbol: Uba7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023738.4 | NP_076227.1 | 5 - 6 | 648 | 57 | |
| GenBank mRNA | AK004894.2 | - | 5 - 6 | 649 | 57 | |
| AK054410.1 | - | 5 - 6 | 655 | 57 | ||
| AK089235.1 | - | 5 - 6 | 652 | 57 | ||
| AK129009.1 | - | 5 - 6 | 560 | 57 | ||
| AK170421.1 | - | 5 - 6 | 641 | 57 | ||
| AK170644.1 | - | 5 - 6 | 647 | 57 | ||
| AK172467.1 | - | 5 - 6 | 610 | 57 | ||
| BC037693.1 | - | 3 - 4 | 422 | 57 | ||
| BC138843.1 | - | 5 - 6 | 565 | 8861865 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uba7
Genomic Map