Assay Details
| Gene Symbol: Tsnaxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024445.4 | NP_077765.1 | 7 - 8 | 831 | 72 | |
| GenBank mRNA | AF343750.1 | - | 7 - 8 | 803 | 72 | |
| AK006041.1 | - | 7 - 8 | 832 | 72 | ||
| BC100431.1 | - | 7 - 8 | 844 | 30917958 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsnaxip1
Genomic Map