Assay Details
| Gene Symbol: Lnx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080795.4 | NP_542985.4 | 7 - 8 | 1751 | 72 | |
| GenBank mRNA | AK036218.1 | - | 7 - 8 | 1752 | 72 | |
| AK047053.1 | - | 7 - 8 | 1735 | 72 | ||
| AK140320.1 | - | 7 - 8 | 1686 | 72 | ||
| AK170146.1 | - | 7 - 8 | 1715 | 72 | ||
| BC090665.1 | - | 7 - 8 | 1717 | 30686384 | 72 | |
| BC117937.1 | - | 7 - 8 | 1706 | 40097802 | 72 | |
| BC117938.1 | - | 7 - 8 | 1705 | 40097810 | 72 | |
More Information
Set Membership: |
|

Lnx2
Genomic Map