Assay Details
| Gene Symbol: Cherp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138585.3 | NP_613051.3 | 12 - 13 | 2291 | 82 | |
| GenBank mRNA | AF536541.1 | - | 12 - 13 | 2193 | 82 | |
| AK049765.1 | - | 12 - 13 | 2202 | 82 | ||
| AK144328.1 | - | 12 - 13 | 2214 | 82 | ||
| AK171907.1 | - | 5 - 6 | 1267 | 82 | ||
| BC032191.1 | - | 12 - 13 | 2209 | 5322208 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cherp
Genomic Map