Assay Details
| Gene Symbol: Cherp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138585.3 | NP_613051.3 | 1 - 2 | 180 | 71 | |
| GenBank mRNA | AF536541.1 | - | 1 - 2 | 82 | 71 | |
| AK049765.1 | - | 1 - 2 | 89 | 71 | ||
| AK144328.1 | - | 1 - 2 | 65 | 71 | ||
| BC032191.1 | - | 1 - 2 | 71 | 5322208 | 71 | |
| BY061354.1 | - | 1 - 2 | 181 | 71 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF536541.1,BC032191.1,NM_138585.3 transcripts.
Set Membership: |
|

Cherp
Genomic Map