Assay Details
| Gene Symbol: Tinf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145705.3 | NP_663751.2 | 6 - 7 | 1250 | 67 | |
| GenBank mRNA | AF518764.1 | - | 6 - 7 | 1215 | 67 | |
| AK011154.1 | - | 6 - 7 | 1218 | 67 | ||
| AK016456.1 | - | 6 - 7 | 1184 | 67 | ||
| AK028952.1 | - | 5 - 6 | 1375 | 67 | ||
| AK090324.1 | - | 6 - 7 | 1217 | 67 | ||
| AK144592.1 | - | 6 - 7 | 1179 | 67 | ||
| BC030347.1 | - | 6 - 7 | 1176 | 5003804 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tinf2
Genomic Map