Assay Details
| Gene Symbol: Cnot10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153585.5 | NP_705813.2 | 13 - 14 | 1862 | 66 | |
| GenBank mRNA | AK029103.1 | - | 13 - 14 | 1737 | 66 | |
| AK149944.1 | - | 13 - 14 | 1692 | 66 | ||
| AK157824.1 | - | 13 - 14 | 1761 | 66 | ||
| AK171701.1 | - | 12 - 13 | 1720 | 66 | ||
| BC025554.1 | - | 13 - 14 | 1723 | 5321869 | 66 | |
| BC036178.1 | - | 10 - 11 | 1318 | 66 | ||
| BC039183.1 | - | 13 - 14 | 1836 | 2631793 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnot10
Genomic Map