Assay Details
| Gene Symbol: Ldlrad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172631.3 | NP_766219.2 | 3 - 4 | 742 | 144 | |
| GenBank mRNA | AK052338.1 | - | 3 - 4 | 743 | 144 | |
| BC096371.1 | - | 2 - 3 | 503 | 3583946 | 144 | |
| BC131933.1 | - | 2 - 3 | 350 | 40130210 | 144 | |
| BC131935.1 | - | 2 - 3 | 350 | 40130212 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ldlrad4
Genomic Map