Assay Details
| Gene Symbol: Agbl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294262.1 | NP_001281191.1 | 3 - 4 | 621 | 72 | |
| NM_174849.3 | NP_777274.1 | 3 - 4 | 510 | 72 | ||
| NR_122116.1 | 3 - 4 | 577 | 72 | |||
| NR_122117.1 | 3 - 4 | 577 | 72 | |||
| GenBank mRNA | AK029676.1 | - | 3 - 4 | 511 | 72 | |
| AK034894.1 | - | 3 - 4 | 599 | 72 | ||
| AK035680.1 | - | 1 - 2 | 78 | 72 | ||
| BC057349.1 | - | 3 - 4 | 511 | 6813901 | 72 | |
| DQ867034.1 | - | 3 - 4 | 506 | 72 | ||
| DQ867035.1 | - | 3 - 4 | 506 | 72 | ||
| DQ867037.1 | - | 3 - 4 | 506 | 72 | ||
| DQ867038.1 | - | 3 - 4 | 506 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agbl5
Genomic Map