Assay Details
| Gene Symbol: Agbl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048192.2 | NP_001041657.1 | 10 - 11 | 1886 | 66 | |
| NM_001294262.1 | NP_001281191.1 | 9 - 10 | 1914 | 66 | ||
| NM_174849.3 | NP_777274.1 | 9 - 10 | 1803 | 66 | ||
| NR_122116.1 | 9 - 10 | 1870 | 66 | |||
| NR_122117.1 | 9 - 10 | 1870 | 66 | |||
| GenBank mRNA | AK029676.1 | - | 9 - 10 | 1804 | 66 | |
| AK034894.1 | - | 9 - 10 | 1892 | 66 | ||
| AK035680.1 | - | 7 - 8 | 1371 | 66 | ||
| AK083656.1 | - | 8 - 9 | 2057 | 66 | ||
| BC057349.1 | - | 9 - 10 | 1804 | 6813901 | 66 | |
| DQ867034.1 | - | 9 - 10 | 1799 | 66 | ||
| DQ867035.1 | - | 9 - 10 | 1799 | 66 | ||
| DQ867036.1 | - | 10 - 11 | 1886 | 66 | ||
| DQ867037.1 | - | 9 - 10 | 1799 | 66 | ||
| DQ867038.1 | - | 9 - 10 | 1799 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agbl5
Genomic Map