Assay Details
| Gene Symbol: Prickle3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290624.1 | NP_001277553.1 | 2 - 3 | 204 | 63 | |
| GenBank mRNA | AK037217.1 | - | 2 - 3 | 167 | 63 | |
| AK079845.1 | - | 2 - 3 | 111 | 63 | ||
| AK083704.1 | - | 1 - 2 | 171 | 63 | ||
| BC023671.1 | - | 2 - 3 | 200 | 63 | ||
| BC049258.1 | - | 2 - 3 | 146 | 5250488 | 63 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK083704.1 transcripts.
Set Membership: |
|

Prickle3
Genomic Map