Assay Details
| Gene Symbol: Bola2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175103.3 | NP_780312.1 | 1 - 2 | 97 | 82 | |
| GenBank mRNA | AK002662.1 | - | 1 - 2 | 100 | 82 | |
| AK003922.1 | - | 1 - 2 | 100 | 82 | ||
| AK028165.1 | - | 1 - 2 | 111 | 82 | ||
| BC045523.1 | - | 1 - 2 | 115 | 6397886 | 82 | |
| BC061197.1 | - | 1 - 2 | 125 | 6707992 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25501817 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25501814 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040362 | AK140279.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040362 | BC045523.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040362 | BC061197.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040362 | NM_175103.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040362 | AK003922.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894335 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302782 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46478 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169701 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040362 | AK140279.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040362 | BC045523.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040362 | BC061197.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040362 | NM_175103.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040362 | AK003922.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25501828 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169701 | - | - |
| Whole Mouse Genome G4122A | A_52_P59817 | AK140279.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P59817 | BC045523.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P59817 | BC061197.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P59817 | NM_175103.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P59817 | AK002662.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104949 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169701 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06152115 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06152122 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06152118 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06152111 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25501814 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25501817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25501875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06106991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06391556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06152111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25501822 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10557454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557451 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557452 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434545_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434544_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455776_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434543_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436639_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10557450 | - | - |
| Mouse Genome 430 2.0 Array | 1434544_at | - | - |
| Mouse Genome 430 2.0 Array | 1434544_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434543_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1434545_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436639_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434545_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1455776_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436639_at | - | - |
| Mouse Genome 430 2.0 Array | 1455776_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1434543_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66162_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66162_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700015457.mm.1 | TC0700001812.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015457.mm.1 | NM_175103.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015457.mm.1 | NM_175103.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015458.mm.1 | TC0700001812.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015456.mm.1 | TC0700001812.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015456.mm.1 | NM_175103.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015456.mm.1 | NM_175103.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015454.mm.1 | TC0700001812.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015454.mm.1 | NM_175103.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015454.mm.1 | NM_175103.3 | Refseq |
| Mouse Expression Set 430 | 1434544_at | - | - |
| Mouse Expression Set 430 | 1434545_PM_x_at | - | - |
| Mouse Expression Set 430 | 1434545_x_at | - | - |
| Mouse Expression Set 430 | 1436639_at | - | - |
| Mouse Expression Set 430 | 1455776_PM_x_at | - | - |
| Mouse Expression Set 430 | 1455776_x_at | - | - |
| Mouse Expression Set 430 | 1434544_PM_at | - | - |
| Mouse Expression Set 430 | 1434543_PM_a_at | - | - |
| Mouse Expression Set 430 | 1436639_PM_at | - | - |
| Mouse Expression Set 430 | 1434543_a_at | - | - |
| Murine Genome U74v2 Set | 104573_at | - | - |
| Murine Genome U74v2 Set | 104573_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436639_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434544_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455776_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434545_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434543_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434544_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455776_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436639_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434543_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434545_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436639_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455776_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434545_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436639_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455776_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434543_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434544_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434544_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434543_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434545_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 918134 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1450022 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1450022 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001450022 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2642537 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2642537 | - | - |
| mouse-6 v1.1 (Illumina) | scl32057.1.564_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1450022 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32057.1.564_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450022 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2846368 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2846368 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32057.1.564_6-S | - | - |

Bola2
Genomic Map