Assay Details
| Gene Symbol: Rps19bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175109.3 | NP_780318.1 | 1 - 2 | 80 | 138 | |
| GenBank mRNA | AB233218.1 | - | 1 - 2 | 76 | 138 | |
| AK010954.1 | - | 1 - 2 | 63 | 139 | ||
| AK075974.1 | - | 1 - 2 | 61 | 138 | ||
| BC038691.1 | - | 1 - 2 | 66 | 1495043 | 138 | |
| BC050115.1 | - | 1 - 2 | 69 | 30017673 | 138 | |
| BY237968.1 | - | 1 - 2 | 80 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps19bp1
Genomic Map