Assay Details
| Gene Symbol: Rae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175112.5 | NP_780321.1 | 1 - 2 | 258 | 138 | |
| GenBank mRNA | AK014325.1 | - | 1 - 2 | 215 | 138 | |
| AK017746.1 | - | 1 - 2 | 179 | 138 | ||
| AK035278.1 | - | 1 - 2 | 204 | 138 | ||
| AK079383.1 | - | 1 - 2 | 239 | 138 | ||
| AK146058.1 | - | 1 - 2 | 223 | 138 | ||
| AK156681.1 | - | 1 - 2 | 223 | 138 | ||
| BC059051.1 | - | 1 - 2 | 180 | 6818771 | 138 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK035278.1,AK079383.1,AK146058.1,AK156681.1,BC059051.1,NM_175112.5 transcripts.
Set Membership: |
|

Rae1
Genomic Map