Assay Details
| Gene Symbol: Zcchc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175126.4 | NP_780335.1 | 1 | 1015 | 72 | |
| GenBank mRNA | AK053859.1 | - | 1 | 752 | 72 | |
| BC055755.1 | - | 1 | 998 | 6412435 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749215 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749220 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749221 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749222 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749219 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749217 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749224 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749218 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749216 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749223 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479132 | BC055755.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479132 | AK053859.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479132 | NM_175126.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479132 | BC055755.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479132 | AK053859.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479132 | NM_175126.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479132 | BC055755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479132 | AK053859.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479132 | NM_175126.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479132 | BC055755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479132 | AK053859.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479132 | NM_175126.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P479132 | BC055755.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P479132 | AK053859.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P479132 | NM_175126.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P479132 | BC055755.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P479132 | AK053859.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P479132 | NM_175126.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749182 | - | - |
| Whole Mouse Genome G4122A | A_51_P479132 | BC055755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479132 | AK053859.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479132 | NM_175126.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479132 | BC055755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479132 | AK053859.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479132 | NM_175126.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106361 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01318125 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01694329 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01694323 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01318127 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749215 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749221 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749217 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749218 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749222 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749223 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749224 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749216 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749220 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02124660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01322405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02124656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01368651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02078261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01472786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01368648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01368649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02124655 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10488628 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488627 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488629 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428401_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428402_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10488626 | - | - |
| Mouse Genome 430 2.0 Array | 1428401_at | - | - |
| Mouse Genome 430 2.0 Array | 1428402_at | - | - |
| Mouse Genome 430 2.0 Array | 1428401_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428402_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67917_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67917_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200041616.mm.1 | NM_175126.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041616.mm.1 | NM_175126.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041616.mm.1 | TC0200004899.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428402_at | - | - |
| Mouse Expression Set 430 | 1428402_PM_at | - | - |
| Mouse Expression Set 430 | 1428401_at | - | - |
| Mouse Expression Set 430 | 1428401_PM_at | - | - |
| Murine Genome U74v2 Set | 103374_PM_at | - | - |
| Murine Genome U74v2 Set | 103375_PM_at | - | - |
| Murine Genome U74v2 Set | 103374_at | - | - |
| Murine Genome U74v2 Set | 103375_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428402_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428401_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428402_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428401_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428401_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428402_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428401_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428402_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 853542 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4780278 | - | - |
| MouseWG-6 v2 Expression BeadChip | 1990035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4780278 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1990035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 580438 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000580438 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001990035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004780278 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2476904 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248465 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2437173 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248465 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2476904 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2437173 | - | - |
| mouse-6 v1.1 (Illumina) | scl067917.1_76-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0067917.1_135-S | - | - |
| mouse-6 v1.1 (Illumina) | scl067917.2_176-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 580438 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4780278 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1990035 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067917.1_135-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067917.2_176-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067917.1_76-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 580438 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780278 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1990035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1248465 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2437173 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2951955 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1248465 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2951955 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2437173 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067917.1_76-S | - | - |

Zcchc3
Genomic Map