Assay Details
| Gene Symbol: 2300002M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175148.3 | NP_780357.1 | 1 - 2 | 140 | 96 | |
| GenBank mRNA | AK009013.1 | - | 1 - 2 | 89 | 96 | |
| AK037046.1 | - | 1 - 2 | 143 | 96 | ||
| BC107266.1 | - | 1 - 2 | 140 | 40052573 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P402144 | AK037046.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P402144 | BC107265.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P402144 | BC107266.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P402144 | NM_175148.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P402144 | AK037046.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P402144 | BC107265.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P402144 | BC107266.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P402144 | NM_175148.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882105 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124851 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P402144 | AK037046.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P402144 | BC107265.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P402144 | BC107266.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P402144 | NM_175148.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P402144 | AK037046.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P402144 | BC107265.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P402144 | BC107266.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P402144 | NM_175148.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159769 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08768571 | - | - |
| Whole Mouse Genome G4122A | A_51_P402144 | AK037046.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402144 | BC107265.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402144 | BC107266.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402144 | NM_175148.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402144 | AK037046.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402144 | BC107265.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402144 | BC107266.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402144 | NM_175148.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112620 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13221174 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13221176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13221170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13221162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159757 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13461677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13221159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13440453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159738 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444889 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429496_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444886 | - | - |
| Mouse Genome 430 2.0 Array | 1429496_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1429496_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69542_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69542_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700007137.mm.1 | NM_175148.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007137.mm.1 | NM_175148.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007137.mm.1 | TC1700000695.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429496_PM_x_at | - | - |
| Mouse Expression Set 430 | 1429496_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429496_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429496_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3190121 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3190121 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003190121 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2683656 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2683656 | - | - |
| mouse-6 v1.1 (Illumina) | scl50771.2.1_175-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3190121 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50771.2.1_175-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190121 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2683656 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2683656 | - | - |

2300002M23Rik
Genomic Map