Assay Details
| Gene Symbol: Tatdn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175151.4 | NP_780360.1 | 1 - 2 | 49 | 139 | |
| GenBank mRNA | AK010232.1 | - | 1 - 2 | 58 | 279 | |
| AK042248.1 | - | 1 - 2 | 50 | 139 | ||
| AK161972.1 | - | 1 - 2 | 58 | 139 | ||
| BC061187.1 | - | 1 - 2 | 46 | 30277245 | 139 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK042248.1,BC061187.1,NM_175151.4 transcripts.
Set Membership: |
|

Tatdn1
Genomic Map