Assay Details
| Gene Symbol: Tatdn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175151.4 | NP_780360.1 | 9 - 10 | 625 | 123 | |
| GenBank mRNA | AK042248.1 | - | 9 - 10 | 626 | 123 | |
| BC034895.1 | - | 6 - 7 | 412 | 123 | ||
| BC061187.1 | - | 9 - 10 | 622 | 30277245 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tatdn1
Genomic Map