Assay Details
| Gene Symbol: Galk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291002.1 | NP_001277931.1 | 5 - 6 | 609 | 58 | |
| NM_175154.3 | NP_780363.1 | 5 - 6 | 715 | 58 | ||
| GenBank mRNA | AK053002.1 | - | 5 - 6 | 565 | 58 | |
| AK082521.1 | - | 5 - 6 | 606 | 58 | ||
| AK152851.1 | - | 3 - 4 | 298 | 58 | ||
| AK154762.1 | - | 5 - 6 | 564 | 58 | ||
| BC079843.1 | - | 5 - 6 | 579 | 5690056 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galk2
Genomic Map