Assay Details
| Gene Symbol: Erich3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175176.3 | NP_780385.1 | 1 - 2 | 198 | 99 | |
| GenBank mRNA | AK076632.1 | - | 1 - 2 | 199 | 99 | |
| BC138236.1 | - | 1 - 2 | 118 | 8861255 | 99 | |
| BC145840.1 | - | 1 - 2 | 118 | 40131143 | 99 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK076632.1,BC138236.1,BC145840.1,NM_175176.3 transcripts.
Set Membership: |
|

Erich3
Genomic Map