Assay Details
| Gene Symbol: Mvb12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175184.4 | NP_780393.2 | 5 - 6 | 1085 | 54 | |
| GenBank mRNA | AK012178.1 | - | 5 - 6 | 578 | 54 | |
| AK087821.1 | - | 5 - 6 | 603 | 54 | ||
| AK131113.1 | - | 5 - 6 | 603 | 54 | ||
| AK135990.1 | - | 5 - 6 | 519 | 54 | ||
| BC049129.1 | - | 5 - 6 | 598 | 6825067 | 54 | |
| BC059907.1 | - | 5 - 6 | 1085 | 6824889 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mvb12b
Genomic Map